n-Pentane 99,9, B&J Brand™ ACS HPLC, Spectrophotometry, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Pentane

312-1L 312-4
CABJ312-1LCS 916.16 CAD
CABJ312-1L CABJ312-4
n-Pentane 99,9, B&J Brand™ ACS HPLC, Spectrophotometry, Burdick & Jackson™
n-Pentane

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36 °C (1.013.25 hPa)
Melting Pt: -130
Density: 0.626 g/cm³ (20 °C)
Flash Pt: 7.8 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC 10 ng/L max.
GC Analysis (n-Pentane) 98 % min.
GC Analysis (total C5 isomers) 99.9 % min.
Refractive Index (20ºC) 1.3573 - 1.3579
Residue 1 mg/L max.
UV Cutoff 190 nm max.
Water by Karl Fischer Titration 0.01 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR