BD Saf-T-Intima™ Closed I.V. Catheter System, BD Medical
Supplier: BD
383335 383336
CABD383335CS 2057.79 CAD
CABD383335 CABD383336
BD Saf-T-Intima™ Closed I.V. Catheter System, BD Medical
Catheters
Sterile, closed I.V
Catheters are made of BD Vialon™ biomaterial. Superior tip integrity, an ultra-smooth microsurface, and an exclusive lubrication process enable easy insertion, making them easier to place and advance on a wider cross-section of patients. Superior softening capabilities permit the catheter to conform to the natural lie of the vessel, reducing pressure on the vessel walls and reducing the risk of penetration of the vessel wall by the catheter material.
Catheters are DEHP-free and do not contain natural rubber latex. Imprinted with lot number.
catheter systems combine advanced needlestick safety with an innovative system design, resulting in virtually bloodless insertion and greater clinician safety. A telescoping needle shield passively covers the stylet as it is withdrawn from the catheter, providing a safeguard from potential sharps injury. The pre-attached extension tubing offers a closed system, ensuring a virtually bloodless venipuncture. Removable PRN adapter allows the use of any needleless closed I.V. access device.
Certifications: CE marked.