Falcon® Cell Scrapers, Sterile, Corning
Supplier: Corning
353085 353086 353089 353087
CA15621-005CS 409.31 CAD
CA15621-005 CA15621-010 CABD353089 CA15621-015
Falcon® Cell Scrapers, Sterile, Corning
Cell Scrapers
PS handle, TPE blade. Designed to provide maximum accessibility to the growth surfaces of a variety of culture vessels.
  • Cross-ribbed PS handle provides greater rigidity, to ensure better control while scraping cells
  • Highly compliant TPE blade pivots to provide multiple angles to remove cells from the entire growth surface
Ordering information: Supplied individually wrapped in peel-open packages.
Order Now
SPECIFICATIONS
Environmentally Preferable
Environmental certification