Digital Stem Thermometers
Supplier: Heathrow Scientific
83110 83210-12
76545-942EA 40.18 CAD
76545-942 76546-706
Digital Stem Thermometers
Thermometers Digital Thermometers
Our digital stem thermometers are ideal for temperature monitoring in essentially any setting. Ideal for easily monitoring reagents and liquids in test tubes, beakers, and flasks, food processing, refrigeration, horticulture and much more.
  • The 83000-series features a dual scale (°C/°F), fast 1-second update time
  • Swiveling head can be moved a full 180° for easy reading in any position
  • Both the ON/OFF switch and the scale selection switch are located on the face for simple access
The thermistor sensor stems are stainless steel, 3.5 mm (0.138") diameter and come with a protective case featuring a pocket clip.
Delivery information: Includes a 1.5 V alkaline coin cell battery (Type LR44) and features automatic power-off after 10 minutes of idle use.