IRS Alcohol Hydrometers
Supplier: Heathrow Scientific
6613-B 6613-F 6613-G 6613-H 6613-I 6613-K 6613-L 6613-M 6613-N 6613-P 6613-Q 6613-R
76546-482EA 89.61 CAD
76546-482 76546-486 76546-490 76546-494 76546-498 76546-502 76546-506 76546-510 76546-514 76546-518 76546-522 76546-526
IRS Alcohol Hydrometers
Hydrometers
These IRS alcohol hydrometers use the proof scale and comply with IRS specifications.
  • Calibration for use at 15.5 °C/60 °F
Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.