TRIonic® Controlled Environment Acid Gloves, Triple-Polymer, AdvanTech, Mapa Professionnel
Supplier: MAPA
517986 517988 517980 518567 518569 518568 518560 517316 518561 517317 517318 517319 517310 517311 517989 517317
32916-100CS 507.56 CAD
32916-100 32916-104 32916-108 32916-229 32916-234 32916-235 32916-237 32916-240 32916-241 32916-250 32916-271 32916-293 32916-318 32916-330 32916-106 CA32916-250 CA32916-271
TRIonic® Controlled Environment Acid Gloves, Triple-Polymer, AdvanTech, Mapa Professionnel
Gloves
Tri-polymer blend gloves offer superior performance to natural rubber gloves when used in wafer fabrication and other applications using aggressive acids and chemical mixtures.
  • Superior resistance to many aggressive chemicals
  • Chlorination process used allows for easy donning, doffing, and double-gloving
  • Precise fit for comfort and reduced hand fatigue
Unique blend of nitrile, neoprene, and natural rubber provides extreme comfort and durability with exceptional chemical and abrasion resistance. Gloves are resistant to etchants, acids, and caustics.More Product Information