Round-Bottom Boiling Flasks, Thermowell, Heavy Wall, Chemglass
Supplier: Chemglass
CG-1515-05 CG-1515-07
80069-098EA 647.67 CAD
80069-098 80069-102
Round-Bottom Boiling Flasks, Thermowell, Heavy Wall, Chemglass
Flasks Round Bottom Flasks
Heavy wall flasks with a round bottom, a [ST] 24/40 outer joint, and a side well to accept a thermometer
Thermometer top is tooled to accept a size 0 rubber stopper.