β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid ≥98%, crystals, ULTRAPURE Bioreagent, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate, 2-Morpholinoethanesulfonic acid hydrate, 2-(N-Morpholino)ethanesulfonic acid monohydrate

Management of Change (MOC) category = N

BAKR4014-01 BAKR4014-02 BAKR4014-04 BAKR4014-07
CAJT4014-1EA 1239.86 CAD
CAJT4014-1 CAJT4014-2 CAJT4014-4 CAJT4014-7
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid ≥98%, crystals, ULTRAPURE Bioreagent, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate

Buffer for liquid chromatography and other molecular biology applications. Lot analysis on label.

Formula: C₆H₁₃NO₄S·1H₂O
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₆H₁₃NO₄S·H₂O) ≥98 %
Appearance Passes Test
Loss on Drying at 130°C 7 - 10 %
Heavy Metals (as Pb) ≤2 ppm
Product Information (not a specification):
pKa at 20 degrees C = 6.15

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR