Molecular sieve A4 (0.4 nm, 4 Å), BAKER ANALYZED®, activated, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = N
BAKR2708-01
BAKR2708-05
CAJT2708-1EA
261.54
CAD
CAJT2708-1
CAJT2708-5
Molecular sieve A4 (0.4 nm, 4 Å), BAKER ANALYZED®, activated, J.T.Baker®
Molecular sieve A4 (0.4 nm, 4 Å)
Pellets. Lot analysis on label.
MDL Number:
MFCD03457537 CAS Number: 70955-01-0 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Loss on Drying at 150°C | ≤5 % |
Bead Size (mm) | 1.6 - 2.5 mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...