Orthophosphoric acid ≥85% (w/w), ARISTAR®, food grade, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Phosphoric acid

404200-22001 3062 404200-03901
CABDH306219LEA 5015.15 CAD
BDH3066-188L CABDH306219L BDH3062-19L
Orthophosphoric acid ≥85% (w/w), ARISTAR®, food grade, VWR Chemicals BDH®
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Melting Pt: 28 °C
Density: 1.71…1.87 g/cm³ (25 °C)
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Assay (H3PO4) 85.0 - 88.0% (W/W)
Arsenic (As) 3 mg/kg max.
Cadmium (Cd) 3 mg/kg max.
Fluoride (F) 10 mg/kg max.
Identification (PO4) To Pass Test
Lead (Pb) 3 mg/kg max.
Manufacture Date Record Actual
Use by Date Record Actual

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR