2-Propanol ≥99.5% ACS, Production Grade, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

BDH2016-55GLM BDH2016-55GLP BDH2016-5GLP BDH2016-1GLP
BDH2016-55GLMEA 7673.6 CAD
BDH2016-55GLM BDH2016-55GLP BDH2016-5GLP BDH2016-1GLP
2-Propanol ≥99.5% ACS, Production Grade, VWR Chemicals BDH®
2-Propanol

Soluble in water. Meets ACS specifications.

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Purity (CH3CHOHCH3) 99.5% Min.
Solubility in Water TO PASS TEST
Carbonyl Compounds: Acetone 0.002% Max.
Carbonyl Compounds: Propionaldehyde 0.002% Max.
Color (APHA) 10 APHA Max.
Residue upon Evaporation 0.001% Max.
Titrable Acid (µeq/g) 0.1 µeq/g Max.
Titrable Base (µeq/g) 0.1 µeq/g Max.
Water 0.2% Max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR