PARA/Gard™ Liquid Paraffin Repellent, TBS®, General Data
Supplier: General Data Healthcare
PG PG-16
CA27900-008CS 1026.24 CAD
CA27900-008 CA27900-010
PARA/Gard™ Liquid Paraffin Repellent, TBS®, General Data
Cleaning Agents Detergents and Surface Cleansers
Liquid spray prevents paraffin from sticking to countertops and equipment, while keeping everything looking brand new.
Pleasant smelling and non-toxic, the spray is safe for metal, wood, or plastic surfaces.
Order Now
SPECIFICATIONS
Environmentally Preferable