2-Propanol ≥70%, Production Grade, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH2014-1GLP BDH2014-55GLP BDH2014-5GLP BDH2014-5GLMS
BDH2014-1GLPCS 970.75 CAD
BDH2014-1GLP BDH2014-55GLP BDH2014-5GLP BDH2014-5GLMS
2-Propanol ≥70%, Production Grade, VWR Chemicals BDH®
2-Propanol

Clear, colorless liquid.

Formula: (CH₃)₂CHOH
Density: 0.8…0.92 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1987
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, Colorless
Specific Gravity at 60°F (0.872 – 0.883) g/mL


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR