Manganese(II) sulfate monohydrate 98-101%, powder, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Manganese sulphate monohydrate, Manganese sulfate monohydrate, Manganous sulfate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate, Manganese(II) sulfate hydrate

Management of Change (MOC) category = N

BAKR2550-01 2550-05
CAJT2550-1CS 1626.74 CAD
CAJT2550-1 JT2550-5 CAJT2550-69
Manganese(II) sulfate monohydrate 98-101%, powder, BAKER ANALYZED® ACS
Manganese (II) sulphate monohydrate

Powder. Lot analysis on label.


  • Lot analysis on label

Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700
Density: 2.95 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9 III
Merck Index: 13,05763

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (MnSO₄·H₂O) 98.0 - 101.0 %
Insoluble Matter ≤0.01 %
Loss on Ignition 10.0 - 12.0 %
pH of 5% Solution at 25°C 2.0 - 4.0
Chloride (Cl) ≤0.005 %
Calcium (Ca) ≤0.005 %
Heavy Metals (as Pb) ≤0.002 %
Iron (Fe) ≤0.002 %
Magnesium (Mg) ≤0.005 %
ACS - Nickel (Ni) ≤0.02 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.05 %
Zinc (Zn) ≤0.005 %
Substances Reducing Permanganate Passes Test
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR