Petroleum spirit, 35…60 °C ≥99.0% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

BDH1145-19L BDH1145-204L BDH1145-4LG
BDH1145-19LEA 1042.11 CAD
BDH1145-19L BDH1145-204L BDH1145-4LG
Petroleum spirit, 35…60 °C ≥99.0% ACS, VWR Chemicals BDH®
Petroleum spirit

Meets ACS specifications for general use.

Boiling Pt: 35…60 °C (1013 hPa)
Density: 0.7
Flash Pt: -10
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear
Water (K.F.) Less than 0.020%
Residue Less than 0.001% (0.0010 g/150 mL)
GC Purity Greater than 99.0% C5 and C6 isomers
n-Pentane (n-C5) Record value
2-Methyl Pentane (n-C6) Record value
Color (APHA) Less than 10
Boiling Range 35 - 60 C
Acidity Passes test (Aqueous layer should not turn blue litmus paper to red.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR