Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
BDH1143-19L
BDH1143-4LG
BDH1143-19LEA
1745.58
CAD
BDH1143-19L
BDH1143-4LG
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
n-Pentane
Clear, colorless liquid.
Formula:
H₃C(CH₂)₃CH₃ Boiling Pt: 35…36 °C (1013 hPa) Melting Pt: –130 °C Density: 0.63 g/cm³ (20 °C) Flash Pt: –40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009498 CAS Number: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3,II REACH: 01-2119459286-30 Merck Index: 13,07193 |
|
Specification Test Results
Appearance | Clear, colorless liquid |
Water (K.F.) | Less than 0.010% |
Residue | Less than 0.001% (0.0010 g/160 mL) |
GC Purity | Greater than 99.9% n-pentane (n-C5) and C5 isomers |
n-Pentane | Greater than 98.0% |
Color (APHA) | Less than 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...