2-Propanol ≥70%, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
BDH1131-5GL
BDH1131-1LP
BDH1131-4LP
BDH1131-204L
BDH1131-5GLEA
551.94
CAD
BDH1131-5GL
BDH1131-1LP
BDH1131-4LP
BDH1131-204L
2-Propanol ≥70%, VWR Chemicals BDH®
2-Propanol
Clear, colorless liquid.
Meets ACS specifications for general use.
Formula:
(CH₃)₂CHOH Density: 0.8…0.92 g/cm³ (20 °C) Flash Pt: 12 Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1987 ADR: 3,II |
|
Specification Test Results
Appearance | Clear, colorless liquid |
Isopropyl Alcohol | 68.0-72.0% |
Water | 28.0-32.0% |
Specific gravity @ 20ºC | 0.863-0.883 |
Color (APHA) | Less than 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...