Magnesium oxide 96.0-100.5% (excluding ignited substances), powder USP, FCC

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Magnesia usta

Management of Change (MOC) category = R

BAKR2480-05 BAKR2480-01
CAJT2480-5EA 881.07 CAD
CAJT2480-5 CAJT2480-1
Magnesium oxide 96.0-100.5% (excluding ignited substances), powder USP, FCC
Magnesium oxide

Light powder.

Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600
Melting Pt: 2800
Density: 3.58 g/cm³ (25 °C)
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now

Specification Test Results

Meets U.S.P Requirements
Food GMP Manufactured Product
Meets F.C.C. Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Assay (MgO) (ignited basis) 96.0 - 100.5 %
USP - Loss on Ignition ≤ 10.0 %
USP - Free Alkali and Soluble Salts ≤ 2.0 %
USP - Acid-Insoluble Substances ≤ 0.1 %
USP - Bulk Density, For Information Only
USP - Calcium (Ca) ≤ 1.1 %
USP - Iron (Fe) ≤ 0.05 %
FCC - Identification Passes Test
FCC - Assay (MgO) (ignited basis) 96.0 - 100.5 %
FCC - Loss on Ignition ≤ 10.0 %
FCC - Free Alkali and Soluble Salts ≤ 2.0 %
FCC - Acid-Insoluble Substances ≤ 0.1 %
FCC - Arsenic (As) ≤ 3 mg/kg
FCC - Calcium Oxide ≤ 1.5 %
FCC - Lead (Pb) ≤ 4 mg/kg
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
STORAGE: Preserve in tight containers.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR