Magnesium chloride hexahydrate 99.0-101.0%, crystals FCC, USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Magnesium dichloride

Management of Change (MOC) category = HR

BAKR2448-05 2448-07 2448-R 2448-01 BAKR2448-07 BAKR2448-R 2448-05 BAKR2448-01
CAJT2448-1CS 2205.57 CAD
CAJT2448-1 JT2448-5 CAJT2448-R JT2448-7 CAJT2448-5 CAJT2448-7 JT2448-R JT2448-1
Magnesium chloride hexahydrate 99.0-101.0%, crystals FCC, USP, J.T.Baker®
Magnesium chloride hexahydrate

  • Food GMP manufactured product

Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
MDL Number: MFCD00149781
CAS Number: 7791-18-6
Merck Index: 13,05684

Order Now

Specification Test Results

Meets F.C.C. Requirements
Meets U.S.P Requirements
Food GMP Manufactured Product
GMP Manufactured Product
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay 99.0 - 101.0 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - pH (1 in 20) 4.5 - 7.0
USP - Insoluble Matter ≤0.005 %
USP - Sulfate (SO₄) ≤0.005 %
USP - Barium (Ba) Passes Test
USP - Calcium (Ca) ≤0.01 %
USP - Potassium (K) Passes Test
FCC - Assay 99.0 - 101.0 %
FCC - Ammonium (NH₄) ≤0.005 %
FCC - Sulfate (SO₄) ≤0.005 %
FCC - Lead (Pb) ≤4 mg/kg
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR