Cyclohexen-1-ylbenzene 97%
Supplier: Thermo Scientific Chemicals
B20602-06
B20602-14
B20602-22
CAAAB20602-06AEA
81.37
CAD
CAAAB20602-06A
CAAAB20602-14A
CAAAB20602-22A
Cyclohexen-1-ylbenzene 97%
Cyclohexen-1-ylbenzene
Liquid
Purity: 97%
CAS No.: 771-98-2
Molecular Formula: C12H14
Formula Weight: 158.24
Flash Point: 103°(217°F)
Refractive Index: 1.5700
Melting Point: -11°
Boiling Point: 250-252°
Beilstein Reference: 1905772
MDL No.: MFCD00001542
Formula:
C₁₂H₁₄ Boiling Pt: 250…252 °C Melting Pt: –11 °C Density: 0.992 Flash Pt: 103 °C(217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001542 CAS Number: 771-98-2 EINECS: 212-242-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...