Magnesium oxide heavy 96,0-100,5 complexometry_ignited basis, EMPROVE® ESSENTIAL E530, BP, Ph. Eur., USP, extra_pure, SAFC®
Supplier: EMD MILLIPORE – PCS CA EXCEP
Synonyms:
Magnesia usta
1.05867.9029
1.05867.1000
CA1.05867.9029EA
2412.42
CAD
CA1.05867.9029
CA1.05867.1000
Magnesium oxide heavy 96,0-100,5 complexometry_ignited basis, EMPROVE® ESSENTIAL E530, BP, Ph. Eur., USP, extra_pure, SAFC®
Magnesium oxide
Heavy white powder. Partially soluble in water. Toxic effects.
Formula:
MgO MW: 40.3 g/mol Boiling Pt: 3600 Melting Pt: 2800 Density: 3.58 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011109 CAS Number: 1309-48-4 EINECS: 215-171-9 Merck Index: 13,05700 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...