L-Phenylalaninol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-(-)-2-Amino-3-phenyl-1-propanol, (S)-(-)-Phenylalaninol, (S)-2-Amino-3-phenylpropan-1-ol, L(-)-2-Amino-3-phenyl-1-propanol, L(-)-Phenylalaninol, H-Phenylalaninol, L(-)-2-Amino-3-phenylpropan-1-ol, (S)-(-)-2-Amino-3-phenylpropan-1-ol

A11586-06 A11586-14
CAAAA11586-06EA 161.16 CAD
CAAAA11586-06 CAAAA11586-14
L-Phenylalaninol 98%
L-Phenylalaninol

Air sensitive.


Purity: 98% CAS No.: 3182-95-4 Molecular Formula: C9H13NO Formula Weight: 151.21 Melting Point: 89-94° Beilstein Reference: 2208238 UN No.: 3259 MDL No.: MFCD00004732

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00004732
CAS Number: 3182-95-4
EINECS: 221-674-4
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR