ASTM Specific Gravity Hydrometers
Supplier: Heathrow Scientific
67111H 67112H 67113H 67114H 67115H 67116H 67117H 67118H 67119H 67120H 67125H 67126H 67127H 67128H 67129H 67130H 67131H 67132H 67133H 67134H
76546-550EA 56.81 CAD
76546-550 76546-552 76546-554 76546-556 76546-558 76546-560 76546-562 76546-564 76546-566 76546-568 76546-570 76546-572 76546-574 76546-576 76546-578 76546-580 76546-582 76546-584 76546-586 76546-588
ASTM Specific Gravity Hydrometers
Hydrometers
Our ASTM specific gravity hydrometers are calibrated at 60 °F and manufactured to the specifications of the American Society for Testing and Materials (ASTM).
  • Calibrated for use at 15.5 °C/60 °F
Select the hydrometer that you need by matching the ASTM number shown. Specific Gravity, also known as relative density, is the ratio of the mass of a liquid to the mass of an equal volume of distilled water.