(±)-1,2-Propanediol ≥99.5% ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

30948-AP 30948-K2 30948-K7 30948-M1
CAAA30948-APEA 87.67 CAD
CAAA30948-AP CAAA30948-K2 CAAA30948-K7 CAAA30948-M1
(±)-1,2-Propanediol ≥99.5% ACS
Propylene glycol

Common Applications: As a substitute for ethylene glycol and glycerol. Good solvent for resins. Dissolves many essential oils, but immiscible with fixed oils.


Purity: 99.5% CAS No.: 57-55-6 Molecular Formula: C3H8O2 Formula Weight: 76.10 Flash Point: 107°(225°F) Physical Form: Liquid Density: 1.036 Refractive Index: 1.4320 Sensitivity: Hygroscopic Odor: Nearly odorless pH: neutral Melting Point: -60° Boiling Point: 186-188° Solubility: Miscible with water, acetone, chloroform. Soluble in ether Beilstein Reference: 1340498 Merck Reference: 14,7855 MDL No.: MFCD00064272


As a substitute for ethylene glycol and glycerol. Good solvent for resins. Dissolves many essential oils, but immiscible with fixed oils.

Formula: CH₃CH(OH)CH₂OH
Boiling Pt: 186…188 °C
Melting Pt: –60 °C
Density: 1.036
Flash Pt: 107 °C (225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
EINECS: 200-338-0
Merck Index: 14,7855
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10
Residue after ignition 0.005%
Titratable acid 0.0005meq/g
Cl 1 ppm
H2O 0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR