Molecular sieve A4 (0.4 nm, 4 Å) Grade 514, beads Mesh 8 - 12, Supelco®

Supplier: MilliporeSigma

Synonyms: Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate

MX1583L-1 MX1583L-4
CAMX1583L-1EA 385 CAD
CAMX1583L-1 CAMX1583L-4
Molecular sieve A4 (0.4 nm, 4 Å) Grade 514, beads Mesh 8 - 12, Supelco®
Molecular sieve A4 (0.4 nm, 4 Å)

Beads.

MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR