N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.8%, DriSolv®, Supelco®

Supplier: MilliporeSigma

Synonyms: 1-Methyl-2-pyrrolidoneNMPMethyl PyrrolidoneN-Methyl-2-pyrrolidinone,NMP1-Methyl-2-pyrrolidinone

Packed in septum-sealed bottle.

MX1396-6 MX1396-6
CAMX1396-6CS 5538.82 CAD
EM-MX1396-6 CAMX1396-6
N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.8%, DriSolv®, Supelco®
N-Methyl-2-pyrrolidone (NMP)

Clear liquid. Free from particulates. For organic synthesis.

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00003193
CAS Number: 872-50-4
Merck Index: 12,06197
https://ca.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg

Order Now

Specification Test Results

Assay (GC) 99.5% min
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Residue after ignition 10ppm max
Water (H2O) 50ppm max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR