Perchloric acid, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®

Supplier: VWR International

10201-VWNQ09 10201-VWNQ03 10201-VWNQ03
CA87003-265EA 1100.28 CAD
87003-265 87003-267 CA87003-265
Perchloric acid, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Perchloric acid

High purity acid for quantitative trace metal analysis at the parts-per-billion (ppb) level.

Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 39 °C (75 hPa)
Melting Pt: –101 °C
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 1873
ADR: 5.1,I
https://ca.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg

Order Now

Specification Test Results

Assay 65 - 71 %
Color max. 10 APHA
Aluminum (Al) max. 1 ppb
Antimony (Sb) max. 0.5 ppb
Arsenic (As) max. 0.5 ppb
Barium (Ba) max. 1 ppb
Beryllium (Be) max. 0.5 ppb
Bismuth (Bi) max. 0.5 ppb
Cadmium (Cd) max. 1 ppb
Calcium (Ca) max. 1 ppb
Cerium (Ce) max. 0.5 ppb
Cesium (Cs) max. 0.5 ppb
Cobalt (Co) max. 0.5 ppb
Copper (Cu) max. 0.5 ppb
Dysprosium (Dy) max. 0.5 ppb
Erbium (Er) max. 0.5 ppb
Europium (Eu) max. 0.5 ppb
Gadolinium (Gd) max. 0.5 ppb
Gallium (Ga) max. 0.5 ppb
Gold (Au) max. 0.5 ppb
Hafnium (Hf) Information Only
Holmium (Ho) max. 0.5 ppb
Indium (In) max. 0.5 ppb
Iron (Fe) max. 1 ppb
Lanthanum (La) max. 0.5 ppb
Lead (Pb) max. 1 ppb
Lithium (Li) max. 0.5 ppb
Lutetium (Lu) max. 0.5 ppb
Magnesium (Mg) max. 1 ppb
Manganese (Mn) max. 1 ppb
Molybdenum (Mo) max. 0.5 ppb
Neodymium (Nd) max. 0.5 ppb
Nickel (Ni) max. 1 ppb
Niobium (Nb) Information Only
Palladium (Pd) max. 0.5 ppb
Platinum (Pt) max. 0.5 ppb
Potassium (K) max. 1 ppb
Praseodymium (Pr) max. 0.5 ppb
Rhodium (Rh) max. 0.5 ppb
Rubidium (Rb) max. 0.5 ppb
Samarium (Sm) max. 0.5 ppb
Scandium (Sc) max. 0.5 ppb
Silver (Ag) max. 1 ppb
Sodium (Na) max. 1 ppb
Strontium (Sr) max. 0.5 ppb
Tantalum (Ta) Information Only
Tellurium (Te) max. 0.5 ppb
Terbium (Tb) max. 0.5 ppb
Thallium (Tl) max. 0.5 ppb
Thorium (Th) max. 1 ppb
Thulium (Tm) max. 0.5 ppb
Tin (Sn) max. 1 ppb
Titanium (Ti) max. 1 ppb
Tungsten (W) Information Only
Uranium (U) max. 0.5 ppb
Vanadium (V) max. 0.5 ppb
Ytterbium (Yb) max. 0.5 ppb
Yttrium (Y) max. 0.5 ppb
Zinc (Zn) max. 1 ppb
Zirconium (Zr) max. 0.5 ppb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR