N-Methyl-2-pyrrolidone (NMP), OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

MX1392-1 MX1392-5 MX1392P-1
CAMX1392-1CS 3990 CAD
CAMX1392-1 CAMX1392-5 CAMX1392P-1
N-Methyl-2-pyrrolidone (NMP), OmniSolv®, Supelco®
N-Methyl-2-pyrrolidone (NMP)

Biosynthesis

Formula: C₅H₉NO
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.7% min
Amines (as dimethylamine) 5ppm max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after ignition 10ppm max
Titrable base 1.0µeq/g max
UV Abs. at 270 nm 1.00AU max
UV Abs. at 280 nm 0.50AU max
UV Abs. at 300 nm 0.10AU max
UV Abs. at 325 nm 0.05AU max
UV Abs. at 350 nm 0.03AU max
UV Abs. at 400 nm 0.005AU max
UV Cut-off 270nm max
Water (H2O) 200ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR