2-Methylbutane ≥95%, Supelco®

Supplier: MilliporeSigma

Synonyms: iso-Pentane

MX0760-1 MX0760-1
CAMX0760-1EA 641.48 CAD
CAMX0760-1 EM-MX0760-1
2-Methylbutane ≥95%, Supelco®
2-Methylbutane

Practical

Formula: CH₃CH₂CH(CH₃)₂
MW: 72.15 g/mol
Boiling Pt: 28 °C (1013 hPa)
Melting Pt: –160 °C
Density: 0.62 g/cm³ (20 °C)
Flash Pt: –51 °C
MDL Number: MFCD00009338
CAS Number: 78-78-4
UN: 1265
ADR: 3,I
https://ca.vwr.com/stibo/web/std.lang.all/04/68/740468.jpg

Order Now

Specification Test Results

Assay (GC) 95% min
Color (APHA) 30 max
Form Clear liquid
Infrared spectrum Conforms to standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR