Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®

Supplier: MilliporeSigma
Danger

MX0488-NP20 MX0488-1 MX0488-6 MX0488P-1 MX0488-1
CAMX0488-36EA 9104.8 CAD
CAMX0488-36 CAMX0488-39 CAMX0488-40 CAMX0488-NP20 CAMX0488-1 CAMX0488-6 CAMX0488P-1 CATXMX0488-1P
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Methanol

Suitable for use in spectrophometry and HPLC and gas chromatography. Gradient controlled for absence of lipophilic interferences at 254 and 235nm. UV cutoff 203nm. Certificate of Analysis on label.

Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Acetone 10ppm max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 250ppt max
Form Clear liquid
Gradient at 235 nm 0.001AU max
Gradient at 254 nm 0.001AU max
Identity (IR-spectrum) Conforms
Residue after evaporation 1ppm max
Titrable acid 0.3µeq/g max
Titrable base 0.2µeq/g max
UV Abs. at 203 nm 1.00AU max
UV Abs. at 205 nm 0.80AU max
UV Abs. at 210 nm 0.25AU max
UV Abs. at 220 nm 0.10AU max
UV Abs. at 230 nm 0.03AU max
UV Abs. at 250 nm 0.01AU max
UV Abs. at 260 nm 0.005AU max
UV Abs. at 300 nm 0.005AU max
UV Cut-off 203nm max
Water (H2O) 0.05% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR