Methanol ≥99.9%, OmniSolv® HR-GC for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

MX0480-1 MX0480-6
CAMX0480-1CS 1359 CAD
CAMX0480-1 CAMX0480-6
Methanol ≥99.9%, OmniSolv® HR-GC for gas chromatography, Supelco®
Methanol

Tested for use in high resolution gas chromatography. Chromatograms supplied with each bottle.

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid, free from particulates
Assay (GC) 99.9% min
Cap. ECD responsive substances as PCNB (peak) 2ng/L max
Cap. ECD responsive substances as PCNB (total) 20ng/L max
Cap. FID responsive substances as decane (peak) 1µg/L max
Cap. FID responsive substances as decane (total) 10µg/L max
Color (APHA) 5 max
Residue after evaporation 1ppm max
Filtered through 0.2 µm filter To pass test
Formaldehyde (HCHO) 1ppm max
Infrared spectrum Conforms to standard
Titrable acid 0.2µeq/g max
Water (H2O) 0.05% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR