D-(-)-Mannitol, GR ACS, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Mannitol(2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

MX0214-1 MX0214-1
CAMX0214-1EA 1139.5 CAD
CAMX0214-1 EM-MX0214-1
D-(-)-Mannitol, GR ACS, Sigma-Aldrich®
D(-)-Mannitol

Meets ACS Specifications

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 164…169 °C
Density: 1.49 g/cm³ (20 °C)
MDL Number: MFCD00064287
CAS Number: 69-65-8
Merck Index: 13,05769
https://ca.vwr.com/stibo/web/std.lang.all/06/13/16640613.jpg

Order Now

Specification Test Results

Color White
Form Powder
Heavy metals (as Pb) 5ppm max
Insoluble matter 0.01% max
Loss on drying (105°C) 0.05% max
Reducing sugars To pass test
Residue after ignition 0.01% max
Specific rotation [a]25/D +23.3-+24.3°
Titrable acid 0.0008meq/g max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR