tert-Butanol ≥99% (by GC), GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol

BX1805-5 BX1805-1 BX1805-4
CABX1805-5EA 1290 CAD
CABX1805-5 CABX1805-1 CABX1805-2 CABX1805-4
tert-Butanol ≥99% (by GC), GR ACS, Supelco®
tert-Butanol

Meets ACS Specifications.

Formula: (CH₃)₃COH
MW: 74.12 g/mol
Boiling Pt: 81…83 °C (1013 hPa)
Melting Pt: 24…25 °C
Density: 0.78 g/cm3 (20 °C)
Flash Pt: 11
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3 II
Merck Index: 12,01577
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.0% min
Carbonyl compounds (as HCHO) 0.01% max
Color (APHA) 20 max
Form Clear liquid (or solid, m.p. 24°C)
Infrared spectrum Conforms to standard
Residue after evaporation 0.003% max
Titrable acid 0.001meq/g max
Water (H2O) 0.1% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR