Methyl ethyl ketone ≥99.5%, OmniSolv®, Supelco®

Supplier: MilliporeSigma
Danger

BX1673-3 BX1673P-1 BX1673P-1
CABX1673-3EA 167 CAD
CABX1673-3 EM-BX1673P-1 CA-BX1673P-1
Methyl ethyl ketone ≥99.5%, OmniSolv®, Supelco®
Methyl ethyl ketone

For spectophotometry and chromatography. UV cutoff 329nm. Lot analysis on label.

Formula: C₂H₅COCH₃
Boiling Pt: 79.6 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.8054 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.5% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 5ppm max
UV Abs. at 329 nm 1.00AU max
UV Abs. at 335 nm 0.30AU max
UV Abs. at 344 nm 0.05AU max
UV Abs. at 350 nm 0.01AU max
UV Abs. at 400 nm 0.005AU max
UV Cut-off 329nm max
Water (H2O) 0.05% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR