Isoamyl alcohol ≥98.5%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

AX1440-3 AX1440-6
CAAX1440-3EA 864 CAD
CAAX1440-3 CAAX1440-6
Isoamyl alcohol ≥98.5%, GR ACS, Supelco®
Isoamyl alcohol

Meets ACS Specifications

Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
Merck Index: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC, corrected for water content) 98.5% min
Acids and esters (as amyl acetate) 0.2% max
Carbonyl compounds (as HCHO) 0.1% max
Color (APHA) 10 max
Form Clear liquid
Residue after evaporation 0.003% max
Titrable acid 0.002meq/g max
Water (H2O) 0.5% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR