β-(N-Morpholino)ethanesufonic acid (MES) monohydrate ≥99.0%, OmniPur®, Millipore®

Supplier: MilliporeSigma

6110-100GM 6120-1KG
CA-EM6110EA 304.07 CAD
CA-EM6110 CA-EM61201KG
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate ≥99.0%, OmniPur®, Millipore®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Formula: C₆H₁₃NO₄S·1H₂O
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929

Order Now

Specification Test Results

pKa at 25°C 5.50–6.50


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR