SP Wilmad-LabGlass Round Bottom Three Vertical Neck Flasks, SP Industries
Supplier: SP Industries
LG-7330-186 LG-7330-194 LG-7330-258 LG-7330-384 LG-7330-370 LG-7330-376 LG-7330-268 LG-7330-248 LG-7330-182
14202-910EA 331.82 CAD
14202-910 14203-116 14203-220 14203-370 14203-440 14203-524 14203-690 14203-886 14203-950 14203-952 14204-102 14204-104 14204-572 14204-694 14204-784 14205-016 14205-208 14205-278 14205-356 14205-376 14205-404 14205-682 14206-280 14206-578 14206-592 14206-630 14206-898 14206-900 14206-908 14206-914 14206-918 14206-922 14207-476 14207-522 14207-530 14207-670 14207-956 14207-966 14207-968 14208-108 14208-220 14208-340 14208-384 14208-406 14208-528 14208-530 14208-610 14208-614 14208-616 14208-852 14208-854 14208-914 14208-920 14209-056 14209-230 14210-006 14210-056 14210-344 14203-442
SP Wilmad-LabGlass Round Bottom Three Vertical Neck Flasks, SP Industries
Flasks Round Bottom Flasks
Available in various sizes, these borosilicate glass round bottom flasks have three vertical necks with Standard Taper outer joints
The side necks are 180° apart