SP Wilmad-LabGlass Round Bottom Vertical Angled Three Neck Flasks, SP Industries
Supplier: SP Industries
ML-1130-700 LG-7331-182 ML-1130-706 ML-1130-712
14202-220EA 204.85 CAD
14202-220 14202-222 14202-224 14202-306 14202-360 14202-850 14202-914 14203-120 14203-224 14203-308 14203-450 14203-694 14203-960 14204-110 14204-284 14204-956 14205-212 14208-224 14208-388 14208-410 14208-538 14208-624 14208-626 14208-628 14209-510 14209-856 14209-936 14209-938 14210-010 14210-060 14210-238 14210-348 14210-528 14210-596 14210-852 14210-664 14210-666
SP Wilmad-LabGlass Round Bottom Vertical Angled Three Neck Flasks, SP Industries
Flasks Round Bottom Flasks
Available in various sizes, these borosilicate glass round bottom flasks have three necks - the center neck is vertical and the side necks are 180° apart and are inclined at an angle of 20° to the vertical
All necks have Standard Taper outer joints.