Methanol ≥99.9%, Purified Plus® ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

PP230-19 PP230-204
CABJPP230-19EA 515.3 CAD
CABJPP230-19 CABJPP230-204
Methanol ≥99.9%, Purified Plus® ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Methanol

Clear liquid. For specialty organic synthesis, preparative scale liquid chromatography, glassware cleaning, and applications requiring ACS reagent-grade solvent. The 54gal. drum is individually analyzed with certificate on label.


  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Density: 0.792 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay by GC Analysis 99.9 % min.
Water by Karl Fischer Titration 0.1 % max.
Carbonyl Compounds 0.001 % max.
Refractive Index (20ºC) 1.3274 - 1.3294
Residue after Evaporation 5 mg/L max.
Solubility in Water Passes test
Substances Darkened by Sulfuric Acid Passes test
Substances Reducing Permanganate Passes test
Titrable Acid 0.3 µeq/g max.
Titrable Base 0.2 µeq/g max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR