Methanol ≥99.8% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

LP230-19 LP230-4 LP230-204 LP230-4P
CABJLP230D19EA 706.89 CAD
CABJLP230D19 CABJLP230-4 BJLP230-204 BJLP230-4P
Methanol ≥99.8% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Methanol

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: CH₃OH
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay by GC Analysis 99.8% min.
Water by Karl Fischer Titration 0.1 % max.
Carbonyl Compounds 0.001% max.
Color (APHA) 10 max.
Residue After Evaporation 0.001% max.
Solubility in Water Passes test
Substances Darkened by Sulfuric Acid Passes test
Substances Reducing Permanganate Passes test
Titrable Acid 0.3 µeq/g max.
Titrable Base 0.2 µeq/g max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR