Methanol ≥99.9%, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Hydroxy methaneMethyl alcoholCarbinolWood alcohol

LC230-4 LC230-2.5 LC230-1 LC230-2.5
CABJLC230-2.5CS 837.84 CAD
CABJLC230-2.5 BJLC230-2.5 BJLC230-4 BJLC230-1
Methanol ≥99.9%, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™
Methanol

HIGH PURITY SOLVENTS, LC-MS GRADE SOLVENTS


  • Extensive quality control to ensure that they meet our specifications
  • Low solvent and metal impurities for reduced interference with delicate, minute or unknown samples
  • Minimal organic and trace metal contamination, which results in higher ionization efficiency, lower adduct formation and easier data interpretation
  • LC-MS tested

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
Merck Index: 13,05984
https://ca.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg

Order Now

Specification Test Results

GC Analysis 99.9 % min.
Water by Karl Fischer Titration 0.05 % max.
HPLC Gradient Elution Passes test
LC-MS Negative Mode (as 4-nitrophenol) 50 ppb max.
LC-MS Positive Mode (as caffeine) 100 ppb max.
LC-MS Positive Mode (as reserpine) 50 ppb max.
Refractive Index (20ºC) 1.3280 - 1.3288
Residue 1 mg/L max.
Trace Metals Passes test
UV Cutoff 205 nm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR