Methanol ≥99.9% ACS, meets analytical specification of USP for HPLC, for spectrophotometry

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

AH230-4PC AH230-4
CABJAH230-4PCCS 981.32 CAD
CABJAH230-4PC CABJAH230-4
Methanol ≥99.9% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Methanol

Meets USP Reagent Specifications


  • ACS certified
  • Lot-to-lot consistency
  • HPLC grade controlled for UV absorbance, GC assay, water content and non-volatile residue

Formula: CH₃OH
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Carbonyl Compounds 0.001 % max.
Color (APHA) 10 max.
GC Analysis 99.9 % min.
HPLC Gradient @ 254 nm 0.005 AU max.
Residue 3 mg/L max.
Titrable Acid 0.3 µeq/g max.
Titrable Base 0.2 µeq/g max.
UV Cutoff 205 nm max.
Water by Karl Fischer Titration 0.05 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR