Nitri Task™ Supported Nitrile Gloves, Honeywell Safety
Supplier: Honeywell Safety
Nitri Task™
NF13/7S NF13/8M NF13/10XL NF13/9L
CA89022-108CS 1247.5 CAD
CA89022-108 CA89022-106 CA89022-104 CA89022-110
Nitri Task™ Supported Nitrile Gloves, Honeywell Safety
Gloves
For use in automotive assembly, general maintenance work, light assembly, manufacturing, and small part handling applications.
  • Knit wrist
  • Color-coded cuff trim assists in easy size identification
  • Minimal shrinkage of nylon shell allows gloves to be laundered
Nitrile palm coating provides excellent grip and durability. Uncoated back design allows gloves to breathe for added comfort.