Methyl ethyl ketone 99,5, B&J Brand™ ACS HPLC, Spectrophotometry, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

247-4 CS247-56
CABJ247-4CS 1600.82 CAD
CABJ247-4 BJCS247-56
Methyl ethyl ketone 99,5, B&J Brand™ ACS HPLC, Spectrophotometry, Burdick & Jackson™
Methyl ethyl ketone

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6
Melting Pt: -86
Density: 0.805 g/cm³ (20 °C)
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3 II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.5 % min.
Refractive Index (20ºC) 1.3776 - 1.3790
Residue 3 mg/L max.
UV Absorbance @ 340 nm 0.1 AU max.
UV Absorbance @ 350 nm 0.02 AU max.
UV Absorbance @ 375 nm 0.01 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 329 nm max.
Water by Karl Fischer Titration 0.05 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR