tert-Butyl methyl ether ≥99.0%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
242-1L
242-4
CABJ242-1LCS
1441.7
CAD
CABJ242-1L
CABJ242-4
tert-Butyl methyl ether ≥99.0%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
tert-Butyl methyl ether
For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.
- Minimal UV contaminants
- High GC purity
- Low water content
- Outstanding lot-to-lot consistency
Formula:
(CH₃)₃COCH₃ Boiling Pt: 55.2 °C Melting Pt: –108.6 °C Density: 0.74 g/cm³ (20 °C) Flash Pt: –27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008812 CAS Number: 1634-04-4 EINECS: 216-653-1 UN: 2398 ADR: 3,II Merck Index: 13,06059 |
|
Specification Test Results
Electron Capture GC | 10 ng/L max. |
GC Analysis | 99 % min. |
Peroxides (as H₂O₂) at time of packaging | 1 ppm max. |
Refractive Index (20ºC) | 1.3680 - 1.3700 |
Residue | 1 mg/L max. |
UV Absorbance @ 225 nm | 0.5 AU max. |
UV Absorbance @ 250 nm | 0.1 AU max. |
UV Cutoff | 210 nm max. |
Water by Karl Fischer Titration | 0.02 % max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...