Methanol ≥99.9%, B&J Brand™ ACS for purge and trap GC analysis, for determination volatile organic compounds, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

232-1L 232-235 232-1L
CABJ232-1LCS 1053.13 CAD
CABJ232-1L CABJ232-235 BJ232-1L
Methanol ≥99.9%, B&J Brand™ ACS for purge and trap GC analysis, for determination volatile organic compounds, Burdick & Jackson™
Methanol

For analysis of trace volatile organic compounds using purge and trap methods.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency


Suitable for EPA Method 8260B.

Formula: CH₃OH
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.3280 - 1.3288
Residue 1 mg/L max.
Volatile Organic Compounds Passes test
Water by Karl Fischer Titration 0.3 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR