Methanol ≥99.8% ACS for production, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH2018-5GLP BDH2018-1GLP
BDH2018-5GLPEA 287.65 CAD
BDH2018-5GLP BDH2018-1GLP
Methanol ≥99.8% ACS for production, VWR Chemicals BDH®
Methanol

Clear liquid. Soluble in water. Meets ACS specifications.

Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear
GC Purity (CH3OH) 99.8% Min.
Solubility in Water Passes Test
Carbonyl Compound (Acetone) 0.001% Max.
Carbonyl Compound (Formaldehyde) 0.001% Max.
Carbonyl Compound (Acetaldehyde) 0.001% Max.
Color (APHA) 10 APHA Max.
Residue upon Evaporation 0.001% Max.
Substances Darkened by Sulfuric Acid Passes Test
Substances Reducing Permanganate Passes Test
Titratable Acid 0.3 μeq/g Max.
Titratable Base 0.2 μeq/g Max.
Water (H2O) 0.1% Max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR