N-Methyl-2-pyrrolidone (NMP) ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH1141-19L BDH1141-4LG BDH1141-4LP
BDH1141-19LEA 1556.14 CAD
BDH1141-19L BDH1141-4LG BDH1141-4LP
N-Methyl-2-pyrrolidone (NMP) ACS, VWR Chemicals BDH®
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear
Water (K.F.) Less than 0.05%
GC Purity Greater than 99.0%
Color (APHA) Less than 50
Chloride Less than 1 ppm (Opalescence must be no greater than that in the standard.)
Free Amines (as CH3NH2) Less than 0.01% (No more than 3.2mL of 0.01N hydrochloric acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR