Methanol ≥99.8% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH1135-1LP BDH1135-204L BDH1135-19L BDH1135-4LG BDH1135-4LP
BDH1135-1LPCS 360.65 CAD
BDH1135-1LP BDH1135-204L BDH1135-19L BDH1135-4LG BDH1135-4LP
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Methanol

Clear liquid. Suitable for histology and cytology applications. Soluble in water.

Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear
Water (K. F.) Less than 0.10%
Residue Less than 0.001% (0.0010 g/125 mL)
GC Purity Greater than 99.8%
Solubility in water Passes test (Solution should be as clear as an equal volume of water)
Color (APHA) Less than 10
Titrable acid Less than 0.3 meq/g (No more than 0.50 mL of 0.01N sodium hydroxide)
Titrable base Less than 0.2 meq/g (No more than 0.40 mL of 0.01N hydrochloric acid)
Carbonyl compounds (as acetone, formaldehyde, and acetaldehyde) Less than 0.001% each
Substances darkened by sulfuric acid Passes test (Solution should be colorless, or have no more color than the acid or methanol before mixing)
Substances reducing permanganate Passes test (Pink color should not be entirely discharged)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR