VITLAB® Round-Bottom Evaporating Flasks, PFA, BrandTech
Supplier: Brandtech
107797 107897 107997
83008-576PK 578.9 CAD
83008-576 83008-578 83008-580
VITLAB® Round-Bottom Evaporating Flasks, PFA, BrandTech
Flasks Round Bottom Flasks
Ideal for use in rotary evaporators and in the distillation of high-purity solvents for trace analysis.
  • Trace analysis grade
  • Easy to clean
  • Max temperature under vacuum 50 °C
Translucent surfaces ensure easy solvent monitoring. Made of PFA (perfluoroalkoxy copolymer). Highly heat- and chemical-resistant, autoclavable, and easy-to-clean. Can withstand temperatures from –200 to 260 °C (–328 to 500 °F). Maximum working temperature under vacuum 50 °C (122 °F). Flasks feature a 29/32 ground socket to fit most rotary evaporators.